Anti-LY9 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 249936]

Supplier: R & D Systems
FAB1898P
220017-263EA 480.12 USD
220017-263
Anti-LY9 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 249936]
Antibodies
Type: Primary
Antigen: LY9
Clonality: Monoclonal
Clone: 249936
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR