Anti-CD9 Mouse Monoclonal Antibody (Fluorescein) [clone: 209306]

Supplier: R & D Systems
FAB1880F
220017-257EA 323.94 USD
220017-257
Anti-CD9 Mouse Monoclonal Antibody (Fluorescein) [clone: 209306]
Antibodies
Type: Primary
Antigen: CD9
Clonality: Monoclonal
Clone: 209306
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR