Anti-ITGA5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 238307]

Supplier: R & D Systems
FAB1864A
220017-255EA 485.91 USD
220017-255
Anti-ITGA5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 238307]
Antibodies
Type: Primary
Antigen: ITGA5
Clonality: Monoclonal
Clone: 238307
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR