Anti-NCR1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 195314]

Supplier: R & D Systems
FAB1850P-100 FAB1850P-025
102881-562EA 513.38 USD
102881-562 102881-560
Anti-NCR1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 195314]
Antibodies
Type: Primary
Antigen: NCR1
Clonality: Monoclonal
Clone: 195314
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR