Anti-NCR1 Mouse Monoclonal Antibody (Fluorescein) [clone: 195314]

Supplier: R & D Systems
FAB1850F-025 FAB1850F-100
102881-556EA 160.52 USD
102881-556 102881-558
Anti-NCR1 Mouse Monoclonal Antibody (Fluorescein) [clone: 195314]
Antibodies
Type: Primary
Antigen: NCR1
Clonality: Monoclonal
Clone: 195314
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR