Anti-NCR3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 210845]

Supplier: R & D Systems
FAB1849A-100 FAB1849A-025
102881-540EA 177.88 USD
102881-540 102881-542
Anti-NCR3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 210845]
Antibodies
Type: Primary
Antigen: NCR3
Clonality: Monoclonal
Clone: 210845
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR