Anti-KIR2DS4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 179315]

Supplier: R & D Systems
FAB1847P-100 FAB1847P-025
102881-538EA 446.86 USD
102881-538 102881-536
Anti-KIR2DS4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 179315]
Antibodies
Type: Primary
Antigen: KIR2DS4
Clonality: Monoclonal
Clone: 179315
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR