Anti-KIR2DS4 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 179315]

Supplier: R & D Systems
FAB1847G-025 FAB1847G-100
102881-534EA 513.38 USD
102881-534 102881-532
Anti-KIR2DS4 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 179315]
Antibodies
Type: Primary
Antigen: KIR2DS4
Clonality: Monoclonal
Clone: 179315
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR