Anti-KIR2DL1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 143211]

Supplier: R & D Systems
FAB1844A-025 FAB1844A-100
102881-508EA 594.37 USD
102881-508 102881-506
Anti-KIR2DL1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 143211]
Antibodies
Type: Primary
Antigen: KIR2DL1
Clonality: Monoclonal
Clone: 143211
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR