Anti-CCR5 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 45531]

Supplier: R & D Systems
FAB182P-100 FAB182P-025
102881-496EA 160.52 USD
102881-496 102881-498
Anti-CCR5 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 45531]
Antibodies
Type: Primary
Antigen: CCR5
Clonality: Monoclonal
Clone: 45531
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR