Anti-CCR5 Mouse Monoclonal Antibody (Fluorescein) [clone: 45531]

Supplier: R & D Systems
FAB182F-025 FAB182F-100
102881-494EA 446.86 USD
102881-494 102881-492
Anti-CCR5 Mouse Monoclonal Antibody (Fluorescein) [clone: 45531]
Antibodies
Type: Primary
Antigen: CCR5
Clonality: Monoclonal
Clone: 45531
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR