Anti-DLL1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 251127]

Supplier: R & D Systems
FAB1818A
220017-225EA 582.8 USD
220017-225
Anti-DLL1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 251127]
Antibodies
Type: Primary
Antigen: DLL1
Clonality: Monoclonal
Clone: 251127
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR