Anti-HAVCR1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 222414]

Supplier: R & D Systems
FAB1817A
102881-488EA 561.11 USD
102881-488
Anti-HAVCR1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 222414]
Antibodies
Type: Primary
Antigen Symbol: HAVCR1
Antigen Name: hepatitis A virus cellular receptor 1
Antigen Synonyms: AI503787, KIM-1, TIM-1, Tim1, Timd1
Clonality: Monoclonal
Clone: 222414
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR