Anti-LILRB3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 222821]

Supplier: R & D Systems
FAB1806P-100 FAB1806P-025
102881-486EA 480.12 USD
102881-486 102881-484
Anti-LILRB3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 222821]
Antibodies
Type: Primary
Antigen: LILRB3
Clonality: Monoclonal
Clone: 222821
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR