Anti-LILRB3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 222821]

Supplier: R & D Systems
FAB1806A-025 FAB1806A-100
102881-482EA 543.75 USD
102881-482 102881-480
Anti-LILRB3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 222821]
Antibodies
Type: Primary
Antigen: LILRB3
Clonality: Monoclonal
Clone: 222821
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR