Anti-CCR5 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: CTC5]

Supplier: R & D Systems
FAB1802P-100 FAB1802P-025
102881-478EA 561.11 USD
102881-478 102881-476
Anti-CCR5 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: CTC5]
Antibodies
Type: Primary
Antigen: CCR5
Clonality: Monoclonal
Clone: CTC5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR