Anti-CCR5 Mouse Monoclonal Antibody (Fluorescein) [clone: CTC5]

Supplier: R & D Systems
FAB1802F-025 FAB1802F-100
102881-474EA 420.83 USD
102881-474 102881-472
Anti-CCR5 Mouse Monoclonal Antibody (Fluorescein) [clone: CTC5]
Antibodies
Type: Primary
Antigen: CCR5
Clonality: Monoclonal
Clone: CTC5
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR