Anti-CCR5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: CTC5]

Supplier: R & D Systems
FAB1802A-025 FAB1802A-100
102881-468EA 202.46 USD
102881-468 102881-470
Anti-CCR5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: CTC5]
Antibodies
Type: Primary
Antigen: CCR5
Clonality: Monoclonal
Clone: CTC5
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR