Anti-CCR9 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 112509]

Supplier: R & D Systems
FAB179C-025 FAB179C-100
102881-442EA 584.25 USD
102881-442 102881-440
Anti-CCR9 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 112509]
Antibodies
Type: Primary
Antigen: CCR9
Clonality: Monoclonal
Clone: 112509
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR