Anti-CCR9 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 112509]

Supplier: R & D Systems
FAB179A-100 FAB179A-025
102881-438EA 608.83 USD
102881-438 102881-436
Anti-CCR9 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 112509]
Antibodies
Type: Primary
Antigen: CCR9
Clonality: Monoclonal
Clone: 112509
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR