Anti-OLR1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 331212]

Supplier: R & D Systems
FAB1798P
102881-434EA 543.75 USD
102881-434
Anti-OLR1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 331212]
Antibodies
Type: Primary
Antigen: OLR1
Clonality: Monoclonal
Clone: 331212
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR