Anti-OLR1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 331212]

Supplier: R & D Systems
FAB1798A
102881-428EA 552.43 USD
102881-428
Anti-OLR1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 331212]
Antibodies
Type: Primary
Antigen: OLR1
Clonality: Monoclonal
Clone: 331212
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR