Anti-MSR1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 268318]

Supplier: R & D Systems
FAB1797P
220017-214EA 513.38 USD
220017-214
Anti-MSR1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 268318]
Antibodies
Type: Primary
Antigen: MSR1
Clonality: Monoclonal
Clone: 268318
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR