Anti-MSR1 Rat Monoclonal Antibody (Fluorescein) [clone: 268318]

Supplier: R & D Systems
FAB1797F
220017-213EA 404.92 USD
220017-213
Anti-MSR1 Rat Monoclonal Antibody (Fluorescein) [clone: 268318]
Antibodies
Type: Primary
Antigen: MSR1
Clonality: Monoclonal
Clone: 268318
Conjugation: Fluorescein
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR