Anti-CCR9 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 248621]

Supplier: R & D Systems
FAB1791A-025 FAB1791A-100
102881-420EA 219.82 USD
102881-420 102881-422
Anti-CCR9 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 248621]
Antibodies
Type: Primary
Antigen: CCR9
Clonality: Monoclonal
Clone: 248621
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR