Anti-ITGB1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 419217]

Supplier: R & D Systems
FAB17783P
102881-360EA 420.83 USD
102881-360
Anti-ITGB1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 419217]
Antibodies
Type: Primary
Antigen: ITGB1
Clonality: Monoclonal
Clone: 419217
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR