Anti-ITGB1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 419127]

Supplier: R & D Systems
FAB17783A
102881-358EA 513.38 USD
102881-358
Anti-ITGB1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 419127]
Antibodies
Type: Primary
Antigen: ITGB1
Clonality: Monoclonal
Clone: 419127
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR