Anti-ITGB1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: P5D2]

Supplier: R & D Systems
FAB17781P
220017-206EA 454.09 USD
220017-206
Anti-ITGB1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: P5D2]
Antibodies
Type: Primary
Antigen: ITGB1
Clonality: Monoclonal
Clone: P5D2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR