Anti-CD83 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HB15e]

Supplier: R & D Systems
FAB1774P
220017-203EA 485.91 USD
220017-203
Anti-CD83 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HB15e]
Antibodies
Type: Primary
Antigen: CD83
Clonality: Monoclonal
Clone: HB15e
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR