Anti-RAET1A Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 186107]

Supplier: R & D Systems
FAB17582A
220017-198EA 577.02 USD
220017-198
Anti-RAET1A Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 186107]
Antibodies
Type: Primary
Antigen: RAET1A
Clonality: Monoclonal
Clone: 186107
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR