Anti-CLEC7A Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 218820]

Supplier: R & D Systems
FAB17561P
220017-197EA 485.91 USD
220017-197
Anti-CLEC7A Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 218820]
Antibodies
Type: Primary
Antigen: CLEC7A
Clonality: Monoclonal
Clone: 218820
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR