Anti-CLEC7A Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 218820]

Supplier: R & D Systems
FAB17561N
102881-342EA 513.38 USD
102881-342
Anti-CLEC7A Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 218820]
Antibodies
Type: Primary
Antigen: CLEC7A
Clonality: Monoclonal
Clone: 218820
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR