Anti-CLEC7A Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 218820]

Supplier: R & D Systems
FAB17561A
220017-196EA 561.11 USD
220017-196
Anti-CLEC7A Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 218820]
Antibodies
Type: Primary
Antigen: CLEC7A
Clonality: Monoclonal
Clone: 218820
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR