Anti-CLEC4A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 216110]

Supplier: R & D Systems
FAB1748A
220017-189EA 577.02 USD
220017-189
Anti-CLEC4A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 216110]
Antibodies
Type: Primary
Antigen: CLEC4A
Clonality: Monoclonal
Clone: 216110
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR