Anti-TNFRSF13B Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 165604]

Supplier: R & D Systems
FAB1741G
102880-942EA 543.75 USD
102880-942
Anti-TNFRSF13B Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 165604]
Antibodies
Type: Primary
Antigen: TNFRSF13B
Clonality: Monoclonal
Clone: 165604
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR