Anti-CXCR4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 44717]

Supplier: R & D Systems
FAB173P-025 FAB173P-100
102880-936EA 189.45 USD
102880-936 102880-938
Anti-CXCR4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 44717]
Antibodies
Type: Primary
Antigen: CXCR4
Clonality: Monoclonal
Clone: 44717
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR