Anti-CXCR4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 44717]

Supplier: R & D Systems
FAB173A-025 FAB173A-100
102881-330EA 566.89 USD
102881-330 102881-328
Anti-CXCR4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 44717]
Antibodies
Type: Primary
Antigen: CXCR4
Clonality: Monoclonal
Clone: 44717
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR