Anti-TREM2 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 237920]

Supplier: R & D Systems
FAB17291N
102881-324EA 600.15 USD
102881-324
Anti-TREM2 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 237920]
Antibodies
Type: Primary
Antigen: TREM2
Clonality: Monoclonal
Clone: 237920
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR