Anti-JAG1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 241002]

Supplier: R & D Systems
FAB1726A
220017-175EA 594.37 USD
220017-175
Anti-JAG1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 241002]
Antibodies
Type: Primary
Antigen: JAG1
Clonality: Monoclonal
Clone: 241002
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR