Anti-CLEC1B Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 219150]

Supplier: R & D Systems
FAB1718A
102881-320EA 577.02 USD
102881-320
Anti-CLEC1B Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 219150]
Antibodies
Type: Primary
Antigen: CLEC1B
Clonality: Monoclonal
Clone: 219150
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR