Anti-CXCR4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 12G5]

Supplier: R & D Systems
FAB170P-100 FAB170P-025
102881-318EA 527.85 USD
102881-318 102881-316
Anti-CXCR4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 12G5]
Antibodies
Type: Primary
Antigen: CXCR4
Clonality: Monoclonal
Clone: 12G5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR