Anti-CXCR4 Mouse Monoclonal Antibody (Fluorescein) [clone: 12G5]

Supplier: R & D Systems
FAB170F-100 FAB170F-025
102881-306EA 389.02 USD
102881-306 102881-304
Anti-CXCR4 Mouse Monoclonal Antibody (Fluorescein) [clone: 12G5]
Antibodies
Type: Primary
Antigen: CXCR4
Clonality: Monoclonal
Clone: 12G5
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR