Anti-CRTAM Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 210213]

Supplier: R & D Systems
FAB16951A
220017-163EA 594.37 USD
220017-163
Anti-CRTAM Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 210213]
Antibodies
Type: Primary
Antigen: CRTAM
Clonality: Monoclonal
Clone: 210213
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR