Anti-IL23R Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 753317]

Supplier: R & D Systems
FAB16861N-100 FAB16861N-025
102881-276EA 513.38 USD
102881-276 102881-274
Anti-IL23R Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 753317]
Antibodies
Type: Primary
Antigen: IL23R
Clonality: Monoclonal
Clone: 753317
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR