Anti-IL23R Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 753317]

Supplier: R & D Systems
FAB16861A-100 FAB16861A-025
102880-920EA 543.75 USD
102880-920 102880-918
Anti-IL23R Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 753317]
Antibodies
Type: Primary
Antigen: IL23R
Clonality: Monoclonal
Clone: 753317
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR