Anti-CXCR3 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 220803]

Supplier: R & D Systems
FAB1685G-025 FAB1685G-100
102880-908EA 527.85 USD
102880-908 102880-906
Anti-CXCR3 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 220803]
Antibodies
Type: Primary
Antigen: CXCR3
Clonality: Monoclonal
Clone: 220803
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR