Anti-CXCR3 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 220803]

Supplier: R & D Systems
FAB1685A-100 FAB1685A-025
102880-898EA 160.52 USD
102880-898 102880-900
Anti-CXCR3 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 220803]
Antibodies
Type: Primary
Antigen: CXCR3
Clonality: Monoclonal
Clone: 220803
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR