Anti-ICOSL Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 136726]

Supplier: R & D Systems
FAB165C
102880-896EA 633.42 USD
102880-896
Anti-ICOSL Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 136726]
Antibodies
Type: Primary
Antigen: ICOSL
Clonality: Monoclonal
Clone: 136726
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR