Anti-PODXL Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 222328]

Supplier: R & D Systems
FAB1658A
102881-264EA 594.37 USD
102881-264
Anti-PODXL Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 222328]
Antibodies
Type: Primary
Antigen: PODXL
Clonality: Monoclonal
Clone: 222328
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR