Anti-SLAMF1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 542301]

Supplier: R & D Systems
FAB1642P
220017-154EA 420.83 USD
220017-154
Anti-SLAMF1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 542301]
Antibodies
Type: Primary
Antigen: SLAMF1
Clonality: Monoclonal
Clone: 542301
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR